4-Butoxyaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

A17528-06 A17528-14 A17528-06 A17528-14
AAAA17528-14EA 132.99 USD
AAAA17528-14 CAAAAA17528-14 CAAAA17528-06 AAA17528-06
4-Butoxyaniline 97%
4-Butoxyaniline
Formula: C₁₀H₁₅NO
Boiling Pt: 84…86 °C/0.05 mm
Density: 0.992
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007866
CAS Number: 4344-55-2
EINECS: 224-402-2
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR