Tetrafluorohydroquinone 98%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 2,3,5,6-TetrafluorohydroquinoneTetrafluorobenzene-1,4-diol

A17088-03 A17088-06 A17088-03 A17088-06
AAA17088-06EA 93.41 USD
AAA17088-06 AAA17088-03 CAAAA17088-06 CAAAA17088-03
Tetrafluorohydroquinone 98%
Tetrafluorohydroquinone
Formula: C₆H₂F₄O₂
MW: 182.07 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00002340
CAS Number: 771-63-1
EINECS: 212-237-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR