Pyridine-2-carboxaldoxime 99%

Warning

Synonyms: 2-Pyridinecarbaldehyde oxime2-Pyridinealdoxime

A16714-14 A16714-22 A16714-14 A16714-22
AAA16714-14EA 80.54 USD
CAAAA16714-14 CAAAA16714-22 AAA16714-14 AAA16714-22
Pyridine-2-carboxaldoxime 99%
Pyridine-2-carboxaldoxime
Formula: C₆H₆N₂O
MW: 122.13 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00006354
CAS Number: 873-69-8
EINECS: 212-849-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR