Cupferron 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium 2-oxo-1-phenylhydrazinolate

A16551-14 A16551-14
AAAA16551-14EA 38.59 USD
AAAA16551-14 CAAAAA16551-14
Cupferron 97+%
Cupferron

Reagent for determination of Ce, Cu, Fe, Sn, and Ti

Formula: C₆H₉N₃O₂
MW: 155.16
Melting Pt: 154 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00078422
CAS Number: 135-20-6
EINECS: 205-183-2
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR