Vitamin A acetate in gelatin, 500,000 I.U./g

Danger

Synonyms: all-trans-Retinyl acetate

A16237-14 A16237-22 A16237-22 A16237-36 A16237-36 A16237-14
AAA16237-22EA 113.46 USD
AAA16237-22 CAAAA16237-36 CAAAA16237-14 AAA16237-14 AAA16237-36 CAAAA16237-22
Vitamin A acetate in gelatin, 500,000 I.U./g
all-trans-Retinyl acetate
Formula: C₂₂H₃₂O₂
MW: 328.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00019413
CAS Number: 127-47-9
EINECS: 204-844-2
Merck Index: 14,10013

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR