Ethyl-2-(trifluoromethyl)benzoate 98%

A16215-14 A16215-06 A16215-14 A16215-22 A16215-22 A16215-06
AAA16215-22EA 1179.93 USD
CAAAA16215-06 AAA16215-22 CAAAA16215-14 CAAAA16215-22 AAA16215-06 AAA16215-14
Ethyl-2-(trifluoromethyl)benzoate 98%
Ethyl 2-(trifluoromethyl)benzoate
Formula: C₁₀H₉F₃O₂
MW: 218.18 g/mol
Boiling Pt: 90…91 °C/10 mm
Density: 1.459
Storage Temperature: Ambient
MDL Number: MFCD00013556
CAS Number: 577-62-8
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR