Decafluorobiphenyl 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Perfluorobiphenyl

A15948-06 A15948-14 A15948-14 A15948-06
AAA15948-06EA 68.69 USD
CAAAA15948-14 CAAAA15948-06 AAA15948-06 AAA15948-14
Decafluorobiphenyl 99%
Decafluorobiphenyl
Formula: C₁₂F₁₀
MW: 334.12 g/mol
Boiling Pt: 206 °C
Melting Pt: 66…70 °C
Density: 1.785
Storage Temperature: Ambient
MDL Number: MFCD00000292
CAS Number: 434-90-2
EINECS: 207-107-3
UN: 3152
ADR: "9",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR