Cyclopentylacetic acid 97+%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 2-Cyclopentylacetic acid

A15696-14 A15696-06 A15696-06 A15696-14 A15696-22 A15696-22
AAA15696-22EA 410 USD
AAA15696-22 CAAAA15696-22 CAAAA15696-06 CAAAA15696-14 AAA15696-06 AAA15696-14
Cyclopentylacetic acid 97+%
Cyclopentylacetic acid
Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 231 °C
Melting Pt: 12…14 °C
Density: 1,023
Flash Pt: 114 °C(237 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001387
CAS Number: 1123-00-8
EINECS: 214-368-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR