4-n-Nonylphenol 98+%

Supplier: Thermo Scientific Chemicals
Danger

A15609-06 A15609-03
AAA15609-06EA 226 USD
AAA15609-06 AAA15609-03
4-n-Nonylphenol 98+%
4-n-Nonylphenol
Formula: C₁₅H₂₄O
MW: 220.35 g/mol
Boiling Pt: 127…130 °C/27 mm
Melting Pt: 43…46 °C
Density: 0.937
Flash Pt: 150 °C(302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002396
CAS Number: 104-40-5
EINECS: 203-199-4
UN: 2430
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR