2-Chloroethyldimethylammonium chloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(2-Chloroethyl)-N,N-Dimethylammonium Chloride2-Dimethylaminoethyl chloride hydrochloride

A15134-22 A15134-22
AAA15134-22EA 29.37 USD
CAAAA15134-22 AAA15134-22
2-Chloroethyldimethylammonium chloride 98+%
2-Chloroethyldimethylammonium chloride
Formula: C₄H₁₁Cl₂N
MW: 144.04 g/mol
Melting Pt: 205…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00012516
CAS Number: 4584-46-7
EINECS: 224-970-1
UN: 3077
ADR: "9",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR