2-Chloroethyldiethylammonium chloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-(2-Chloroethyl)-N,N-diethylammonium chloride2-Diethylaminoethylchloride hydrochloride

A15107-0E A15107-0E
AAA15107-0EEA 222.99 USD
AAA15107-0E CAAAA15107-0E
2-Chloroethyldiethylammonium chloride 98+%
2-Chloroethyldiethylammonium chloride
Formula: C₆H₁₅Cl₂N
MW: 172.1 g/mol
Melting Pt: 206…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00012519
CAS Number: 869-24-9
EINECS: 212-786-4
UN: 2928
ADR: "6.1",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR