Pyridine-2,4-dicarboxylic acid, (max. 10% H₂O) 98%

Synonyms: 2,4-Pyridinedicarboxylic acid monohydrate Lutidinic acid monohydrate

A14409-14 A14409-06 A14409-14 A14409-22 A14409-06 A14409-22
AAA14409-06EA 71.07 USD
AAA14409-06 AAA14409-14 CAAAA14409-06 AAAA14409-22 CAAAAA14409-22 CAAAA14409-14
Pyridine-2,4-dicarboxylic acid, (max. 10% H₂O) 98%
Pyridine-2,4-dicarboxylic acid
Formula: C₇H₅NO₄·1H₂O
MW: 167.12 g/mol
Melting Pt: ca 245 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149413
CAS Number: 207671-42-9
EINECS: 207-892-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR