Tetramethylenebis(triphenylphosphonium bromide) 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14214-18 A14214-09 A14214-09 A14214-18
AAA14214-18EA 184.37 USD
CAAAA14214-09 AAA14214-18 CAAAA14214-18 AAA14214-09
Tetramethylenebis(triphenylphosphonium bromide) 98+%
Tetramethylenebis(triphenylphosphonium bromide)
Formula: C₄₀H₃₈Br₂P₂
Melting Pt: 292…293 °C
Storage Temperature: Ambient
MDL Number: MFCD00031714
CAS Number: 15546-42-6
EINECS: 239-598-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR