1-Benzyl-3-pyrrolidinone 98%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 1-(Phenylmethyl)-3-pyrrolidinone1-Benzylpyrrolidin-3-one

A13426-03 A13426-14 A13426-06 A13426-03 A13426-06 A13426-14
AAAA13426-06EA 153 USD
AAAA13426-06 CAAAAA13426-06 AAAA13426-03 AAAA13426-14 CAAAAA13426-14 CAAAAA13426-03
1-Benzyl-3-pyrrolidinone 98%
1-Benzyl-3-pyrrolidinone
Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Boiling Pt: 77…78 °C/0.01 mm
Density: 1,091
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005342
CAS Number: 775-16-6
EINECS: 212-274-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR