trans-4-Methoxycinnamaldehyde 98+%
|
Warning
|
Synonyms:
(E)-3-(4-Methoxyphenyl)acrylaldehyde
A12839-09
A12839-18
A12839-09
A12839-18
AAA12839-18EA
261.53
USD
AAA12839-18
CAAAA12839-09
AAA12839-09
CAAAA12839-18
trans-4-Methoxycinnamaldehyde 98+%
trans-4-Methoxycinnamaldehyde
Formula:
C₁₀H₁₀O₂ MW: 162.19 Boiling Pt: 160 °C/3 mm Melting Pt: 56…60 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00017343 CAS Number: 24680-50-0 EINECS: 217-807-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...