p-Toluic hydrazide 98%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 4-Methylbenzohydrazidep-Toluoylhydrazinep-Toluic acid hydrazidep-Toluic Hydrazide

A12604-14 A12604-14
AAA12604-14EA 83.6 USD
AAA12604-14 CAAAA12604-14
p-Toluic hydrazide 98%
p-Toluic hydrazide
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00007607
CAS Number: 3619-22-5
EINECS: 222-815-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR