Dihexyl phthalate 97%

Danger

A12593-30 A12593-09 A12593-18 A12593-09 A12593-30 A12593-18
AAA12593-18EA 282.92 USD
CAAAA12593-09 CAAAA12593-18 AAA12593-18 CAAAA12593-30 AAA12593-30 AAA12593-09
Dihexyl phthalate 97%
Dihexyl phthalate
Formula: C₂₀H₃₀O₄
Boiling Pt: 185…187 °C/0.5 mm
Density: 1.01
Storage Temperature: Ambient
MDL Number: MFCD00043720
CAS Number: 84-75-3
EINECS: 201-559-5
UN: 3082
ADR: "9",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR