Ethyl-3,5-dichloro-4-hydroxybenzoate 99%

Synonyms: Ethyl 3,5-dichloro-4-hydroxybenzoate

A12489-14 A12489-22 A12489-06 A12489-06 A12489-14 A12489-22
AAAA12489-06EA 43.37 USD
AAAA12489-06 CAAAAA12489-06 CAAAA12489-22 CAAAA12489-14 AAA12489-14 AAA12489-22
Ethyl-3,5-dichloro-4-hydroxybenzoate 99%
Ethyl 3,5-dichloro-4-hydroxybenzoate
Formula: C₉H₈Cl₂O₃
MW: 235.07 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00016420
CAS Number: 17302-82-8
EINECS: 241-331-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR