Methyl-3-chloro-4-hydroxybenzoate 98%

Warning

Synonyms: Methyl 3-chloro-4-hydroxybenzoate

A12389-22 A12389-22 A12389-14 A12389-14 A12389-06
AAA12389-14EA 259.97 USD
AAA12389-14 AAA12389-22 CAAAA12389-14 CAAAA12389-22 AAAA12389-06
Methyl-3-chloro-4-hydroxybenzoate 98%
Methyl 3-chloro-4-hydroxybenzoate
Formula: C₈H₇ClO₃
MW: 186.59 g/mol
Melting Pt: 104…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00016418
CAS Number: 3964-57-6
EINECS: 223-573-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR