4-(4-Chlorophenyl)piperidin-4-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(4-Chlorophenyl)-4-hydroxypiperidine

A12137-06 A12137-06 A12137-03 A12137-03
AAA12137-03EA 31.79 USD
CAAAA12137-06 AAA12137-03 CAAAA12137-03 AAA12137-06
4-(4-Chlorophenyl)piperidin-4-ol 97%
4-(4-Chlorophenyl)piperidin-4-ol
Formula: C₁₁H₁₄ClNO
MW: 211.69 g/mol
Melting Pt: 137…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00006001
CAS Number: 39512-49-7
EINECS: 254-479-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR