Sodium benzenesulfinate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Benzenesulphinic acid sodium saltBenzenesulfinic acid sodium saltBenzenesulphinic acid, sodium saltBenzenesulfinic acid, sodium salt

A12118-14 A12118-14 A12118-36 A12118-36 A12118-22 A12118-22
AAA12118-22EA 70.79 USD
CAAAA12118-22 CAAAA12118-14 CAAAA12118-36 AAA12118-22 AAA12118-14 AAA12118-36
Sodium benzenesulfinate 98%
Sodium benzenesulphinate
Formula: C₆H₅NaO₂S
MW: 164.16 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013135
CAS Number: 873-55-2
EINECS: 212-842-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR