(2-Chlorophenoxy)acetonitrile 97%

Warning

A11346-14 A11346-22 A11346-14 A11346-06 A11346-06 A11346-22
AAA11346-06EA 71.93 USD
AAA11346-06 AAA11346-14 CAAAA11346-06 AAA11346-22 CAAAA11346-14 CAAAA11346-22
(2-Chlorophenoxy)acetonitrile 97%
(2-Chlorophenoxy)acetonitrile
Formula: C₈H₆ClNO
MW: 167.60
Boiling Pt: 109…110 °C/1 mm
Storage Temperature: Ambient
MDL Number: MFCD00017331
CAS Number: 43111-31-5
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR