4-(4-Nitrobenzyloxy)benzaldehyde 98%

A11067-22 A11067-14 A11067-14 A11067-22 A11067-06 A11067-06
AAAA11067-22EA 734.82 USD
CAAAA11067-14 CAAAAA11067-22 AAAA11067-22 CAAAA11067-06 AAA11067-14 AAA11067-06
4-(4-Nitrobenzyloxy)benzaldehyde 98%
4-(4-Nitrobenzyloxy)benzaldehyde
Formula: C₁₄H₁₁NO₄
MW: 257.25 g/mol
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00017041
CAS Number: 67565-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR