(2-Methylbenzyl)triphenylphosphonium chloride 98+%

Warning

A10719-18 A10719-09 A10719-30 A10719-30 A10719-18 A10719-09
AAA10719-09EA 25.24 USD
CAAAA10719-09 CAAAA10719-30 AAA10719-09 AAA10719-18 AAA10719-30 CAAAA10719-18
(2-Methylbenzyl)triphenylphosphonium chloride 98+%
(2-Methylbenzyl)triphenylphosphonium chloride
Formula: C₂₆H₂₄ClP
Melting Pt: 270…275 °C
Storage Temperature: Ambient
MDL Number: MFCD00031577
CAS Number: 63368-36-5
EINECS: 264-112-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR