Phenylarsonic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

A10625-22 A10625-22
AAA10625-22EA 465.99 USD
CAAAA10625-22 AAA10625-22
Phenylarsonic acid 97%
Phenylarsonic acid
Formula: C₆H₇AsO₃
MW: 202.04 g/mol
Melting Pt: 158…162 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002097
CAS Number: 98-05-5
EINECS: 202-631-9
UN: 3465
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR