4-Acetamido-3-nitrobenzoic acid 97+%

Warning

A10360-06 A10360-14 A10360-06 A10360-22 A10360-14 A10360-22
AAA10360-14EA 172.36 USD
CAAAA10360-06 AAA10360-14 AAAA10360-22 CAAAAA10360-22 AAA10360-06 CAAAA10360-14
4-Acetamido-3-nitrobenzoic acid 97+%
4-Acetamido-3-nitrobenzoic acid
Formula: C₉H₈N₂O₅
Melting Pt: 223…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00014703
CAS Number: 1539-06-6
EINECS: 216-265-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR