Ethyl-5-bromo-2-chlorobenzoate 98+%

Warning

Synonyms: Ethyl 5-bromo-2-chlorobenzoate

A10334-22 A10334-22 A10334-06 A10334-14 A10334-06 A10334-14
AAA10334-22EA 344.1 USD
CAAAA10334-14 AAA10334-22 CAAAA10334-22 AAA10334-06 AAA10334-14 CAAAA10334-06
Ethyl-5-bromo-2-chlorobenzoate 98+%
Ethyl 5-bromo-2-chlorobenzoate
Formula: C₉H₈BrClO₂
MW: 263.52 g/mol
Boiling Pt: 156…157 °C/13 mm
Density: 1.480
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013635
CAS Number: 76008-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR