Benzoin methyl ether 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methoxy-1,2-diphenylethanone1,2-Diphenyl-2-methoxyethan-1-one

A10296-36 A10296-14 A10296-22 A10296-22 A10296-36 A10296-14
AAA10296-22EA 161.99 USD
CAAAA10296-22 CAAAA10296-36 CAAAA10296-14 AAA10296-22 AAA10296-36 AAA10296-14
Benzoin methyl ether 97%
Benzoin methyl ether
Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Boiling Pt: 188…189 °C/15 mm
Melting Pt: 46…50 °C
Density: 1.128
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008492
CAS Number: 3524-62-7
EINECS: 222-538-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR