ALFA88697
88697-18 88697-09 88697-03
CAAA88697-09 CAAA88697-18 CAAA88697-03
ALFA88697
For reducing organic nitro compounds, used in manufacture of sulfur dyes, in ore flotation, and used in preparation of metal sulfide finishes
Soluble in water and alcohol