Neodymium ≥99.9% (REO, rare earth oxide basis), powder -200 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
045914-14
045914-06
AA45914-06EA
139
USD
AA45914-06
AA45914-14
Neodymium ≥99.9% (REO, rare earth oxide basis), powder -200 mesh
Neodymium
Formula:
Nd Boiling Pt: 2474 °C (1013 hPa) Melting Pt: 1014.85 °C Density: 6.46 g/cm³ (1240 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011130 CAS Number: 7440-00-8 EINECS: 231-109-3 UN: 1383 ADR: 4.2,I Merck Index: 12,06538 |
Specification Test Results
Form | Packaged under argon |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...