Boron nitride, bar, Length 300 mm, Width 12.7 mm, Height 12.7 mm

45721-KF
AA45721-KFEA 498.73 USD
AA45721-KF
Boron nitride, bar, Length 300 mm, Width 12.7 mm, Height 12.7 mm
Boron nitride
Formula: BN
MW: 24.82 g/mol
Melting Pt: 2527 °C
Density: 2.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011317
CAS Number: 10043-11-5
EINECS: 233-136-6
Merck Index: 13,01336

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR