Tetrahexylammonium hydrogen sulfate 98%

Supplier: Thermo Scientific Chemicals
Warning

43882-04 43882-04
AA43882-04EA 99.69 USD
CAAA43882-04 AA43882-04
Tetrahexylammonium hydrogen sulfate 98%
Tetrahexylammonium hydrogen sulfate
Formula: C₂₄H₅₃NO₄S
Melting Pt: 96…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00037675
CAS Number: 32503-34-7
EINECS: 251-069-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR