Triethylaluminium ≥94%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Triethylaluminum

43243-22 43243-22
AA43243-22EA 398.99 USD
AA43243-22 CAAA43243-22
Triethylaluminium ≥94%
Triethylaluminium

Catalyst for polymerization of olefins, intermediate
Pyrophoric. Packaged in non-returnable cylinders


Catalyst for polymerization of olefins, intermediate

Formula: C₆H₁₅Al
MW: 114.17 g/mol
Boiling Pt: 128…130 °C (50 mmHg)
Melting Pt: –50 °C
Density: 0.835
Flash Pt: 18 °C (–1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009015
CAS Number: 97-93-8
EINECS: 202-619-3
UN: 3394
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR