Cadmium(II) acetylacetonate 98%

Warning

Synonyms: Cadmium 2,4-pentanedionateCadmium acetylacetonate

41968-14 41968-14 41968-06 41968-22 41968-06
AA41968-14EA 153.94 USD
CAAA41968-22 CAAA41968-06 AA41968-14 CAAA41968-14 AA41968-06
Cadmium(II) acetylacetonate 98%
Cadmium (II) acetylacetonate
Formula: C₁₀H₁₄CdO₄
MW: 310.63 g/mol
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00040412
CAS Number: 14689-45-3
EINECS: 238-730-9
UN: 2570
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR