3-(Trimethylsilyl)-2-oxazolidinone

Warning

Synonyms: 3-(Trimethylsilyl)oxazolidin-2-one

41209-14 41209-03 41209-06 41209-06 41209-14 41209-03
AA41209-06EA 50.13 USD
AA41209-06 CAAA41209-14 CAAA41209-03 CAAA41209-06 AA41209-03 AA41209-14
3-(Trimethylsilyl)-2-oxazolidinone
3-(Trimethylsilyl)-2-oxazolidinone
Formula: C₆H₁₃NO₂Si
MW: 159.26 g/mol
Boiling Pt: 99…100 °C (6 mmHg)
Density: 1,013
Flash Pt: 92 °C (198 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010197
CAS Number: 43112-38-5
EINECS: 256-098-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR