Potassium hexachlororhodate (III) ≥99.99% (metals basis) Rh ≥ 23.3%, Premion®

Supplier: Alfa Aesar
Warning

41013-06 41013-03 41013-03 41013-01 41013-01 41013-06
AA41013-06EA 726.28 USD
AA41013-06 CAAA41013-03 AA41013-03 AA41013-01 CAAA41013-01 CAAA41013-06
Potassium hexachlororhodate (III) ≥99.99% (metals basis) Rh ≥ 23.3%, Premion®
Potassium hexachlororhodate (III)
Formula: Cl₆K₃Rh
MW: 432.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00192414
CAS Number: 13845-07-3
EINECS: 237-569-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR