Diethyl methylphosphonite

Supplier: Thermo Scientific Chemicals

Warning

30357-02 30357-04
AA30357-04EA 287 USD
AA30357-04 AA30357-02
Diethyl methylphosphonite
Diethyl methylphosphonite
Formula: C₅H₁₃O₂P
Storage Temperature: Ambient
MDL Number: MFCD00009089
CAS Number: 15715-41-0
EINECS: 239-805-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR