Chloro(dimethyl)phosphine 97%

Danger

Synonyms: Dimethylchlorophosphine

030160-02 30160-02
AA30160-02EA 196.99 USD
CAAA30160-02 AA30160-02
Chloro(dimethyl)phosphine 97%
Chloro(dimethyl)phosphine
Formula: C₂H₆ClP
MW: 96.5 g/mol
Boiling Pt: 76…77 °C
Density: 1.22
Flash Pt: –18 °C (–1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013620
CAS Number: 811-62-1
EINECS: 212-372-3
UN: 2845
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR