Dichloro(propyl)phosphine 98%

Danger

Synonyms: n-Propyldichlorophosphine

18329-06 18329-03 18329-03 18329-06
AA18329-03EA 46.88 USD
CAAA18329-06 AA18329-03 CAAA18329-03 AA18329-06
Dichloro(propyl)phosphine 98%
Dichloro(propyl)phosphine
Formula: C₃H₇Cl₂P
MW: 144.97 g/mol
Melting Pt: 135…136 °C
Density: 1.118
Storage Temperature: Ambient
MDL Number: MFCD00013618
CAS Number: 15573-31-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR