Anti-CD247 (TCR z chain) Mouse Monoclonal Antibody [clone: 6B10.2]

Supplier: BioLegend
644101 644102
644101-BLEA 147.52 USD
644101-BL 644102-BL
Anti-CD247 (TCR z chain) Mouse Monoclonal Antibody [clone: 6B10.2]
Antibodies
Purified anti-CD247 (TCRζ, CD3ζ) [6B10.2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 0.5-1 μg per ml antibody dilution buffer. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD247 (TCR z chain)
Clonality: Monoclonal
Clone: 6B10.2
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR