Anti-PROX1 Mouse Monoclonal Antibody [clone: 5G10]

Supplier: Rockland Immunochemicals
200-301-E18
RL200-301-E18EA 528.48 USD
RL200-301-E18
Anti-PROX1 Mouse Monoclonal Antibody [clone: 5G10]
Antibodies
Anti-Prox1 (Mouse) antibody is suitable for use in IHC. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Immunochemistry: 1:1000

Type: Primary
Antigen: PROX1
Clonality: Monoclonal
Clone: 5G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Chicken, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR