Proteinase K
Supplier: Diagnostic BioSystems
K030 K030
99990-090EA 151.53 USD
99990-090 CA99990-090
Proteinase K
Enzymes
Proteinase K is specifically formulated to achieve proteolytic digestion of formalin fixed, paraffin embedded tissue sections prior to performing in situ hybridization procedures.