Anti-Protein A Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-101-077S 200-101-077
RL200-101-077SEA 147.5 USD
RL200-101-077S RL200-101-077
Anti-Protein A Goat Polyclonal Antibody
Antibodies
Suitable for immunoprecipitation, immunodiffusion, conjugation and most immunological methods requiring high titer and specificity.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Tag
Antigen: Protein A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR