Anti-PKA RI Rabbit Polyclonal Antibody

A00947-40UG
95045-006EA 119.82 USD
95045-006
Anti-PKA RI Rabbit Polyclonal Antibody
Type: Primary
Antigen: PKA RI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR