Anti-PIN1 Rabbit Polyclonal Antibody

A01090-40UG
95045-676EA 119.78 USD
95045-676
Anti-PIN1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: PIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR