Anti-integrin b7 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: FIB27]

Supplier: BioLegend
121008 121007
121008-BLEA 348.72 USD
121008-BL 121007-BL
Anti-integrin b7 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: FIB27]
Antibodies
PerCP/Cy5.5 anti-human/mouse integrin β7 [FIB27]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application. <br />* PerCP/Cy5.5 has a maximum absorption of 482 nm and a maximum emission of 690 nm.

Type: Primary
Antigen: integrin b7
Clonality: Monoclonal
Clone: FIB27
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR