Anti-PEN2 Rabbit Polyclonal Antibody

A00882-40UG
95044-154EA 119.82 USD
95044-154
Anti-PEN2 Rabbit Polyclonal Antibody
Type: Primary
Antigen: PEN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR