Anti-CD127 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: A7R34]

Supplier: BioLegend
135016 135015
135015-BLEA 158.69 USD
135015-BL 135016-BL
Anti-CD127 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: A7R34]
Antibodies
PE/Cy5 anti-mouse CD127 (IL-7Rα) [A7R34]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.15 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD127
Clonality: Monoclonal
Clone: A7R34
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR