Anti-lacZ Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-4136S
RL200-4136SEA 145.12 USD
RL200-4136S
Anti-lacZ Rabbit Polyclonal Antibody
Antibodies
Suitable for immunoblotting (western or dot blot), ELISA, immunofluorescence microscopy, immunoprecipitation, conjugation and most immunological methods requiring high titer and specificity.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:5000 - 1:10000; Immunochemistry: 1:1500;

Type: Tag
Antigen: lacZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR