Anti-ATM Mouse Monoclonal Antibody [clone: 10H11.E12]

Supplier: Rockland Immunochemicals
200-301-400S
RL200-301-400SEA 181.76 USD
RL200-301-400S
Anti-ATM Mouse Monoclonal Antibody [clone: 10H11.E12]
Antibodies
Protein A Purified Mab anti-ATM has been tested by ELISA and western blotting against both the native and recombinant forms of the Protein. The antibody immunoprecipitates ATM from irradiated human and mouse cells.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Immunofluorescence: 1:100 - 1:500; Western Blot: 1:200 - 1:2000;

Type: Primary
Antigen: ATM
Clonality: Monoclonal
Clone: 10H11.E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR